Copper standard solution, for ICP, in 5% HNO₃, VHG
Supplier: LGC Standards PROMOCHEM
VHG-PCUN-100 VHG-PCUN-500 VHG-PCUN-250 VHG-PCUN-50 VHG-TCUN-100 VHG-TCUN-500
PROOVHG-PCUN-100EA 63.5 EUR
PROOVHG-PCUN-100 PROOVHG-PCUN-500 PROOVHG-PCUN-250 PROOVHG-PCUN-50 PROOVHG-TCUN-100 PROOVHG-TCUN-500
Copper standard solution, for ICP, in 5% HNO₃, VHG
Standards Cationic Standards Copper Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.