Vanadium standard solution, for ICP, in HNO₃, CRM, VHG
Supplier: LGC Standards PROMOCHEM
VHG-PVN-100 VHG-LVN-100 VHG-PVN-50 VHG-PVN-500 VHG-TVN-100 VHG-TVN-500
PROOVHG-PVN-100EA 63.5 EUR
PROOVHG-PVN-100 PROOVHG-LVN-100 PROOVHG-PVN-50 PROOVHG-PVN-500 PROOVHG-TVN-100 PROOVHG-TVN-500
Vanadium standard solution, for ICP, in HNO₃, CRM, VHG
Standards Cationic Standards Vanadium Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.