Total organic carbon standards, VHG
Supplier: LGC Standards PROMOCHEM
VHG-TOC1K-100
PROOVHG-TOC1K-100EA 77.5 EUR
PROOVHG-TOC1K-100
Total organic carbon standards, VHG
Standards Environmental Analysis Standards
This standard solution is intended for use as a calibration and reference standard for total organic carbon (TOC).
Certifications: Accredited to ISO 9001 and ISO/IEC 17025.