Tungsten standard solution, for ICP, in 5% HNO₃, tr. HF, CRM, VHG
Supplier: LGC Standards PROMOCHEM
VHG-PWNF-100 VHG-PWNF-500 VHG-TWNF-100 VHG-TWNF-500
PROOVHG-PWNF-100EA 64.8 EUR
PROOVHG-PWNF-100 PROOVHG-PWNF-500 PROOVHG-TWNF-100 PROOVHG-TWNF-500
Tungsten standard solution, for ICP, in 5% HNO₃, tr. HF, CRM, VHG
Standards Cationic Standards Tungsten Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.
Order Now
SPECIFICATIONS
Environmentally Preferable