Anti-EEF1G Rabbit Polyclonal Antibody
Supplier: ProSci Inc.
30-371
PRSI30-371EA 660 EUR
PRSI30-371
Anti-EEF1G Rabbit Polyclonal Antibody
Antibodies
EEF1G is a subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This subunit contains an N-terminal glutathione transferase domain, which may be involved in regulating the assembly of multisubunit complexes containing this elongation factor and aminoacyl-tRNA synthetases.This gene encodes a subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This subunit contains an N-terminal glutathione transferase domain, which may be involved in regulatingMore Product Information