Anti-CDY1 Rabbit Polyclonal Antibody
Supplier: ProSci Inc.
30-429
PRSI30-429EA 601 EUR
PRSI30-429
Anti-CDY1 Rabbit Polyclonal Antibody
Antibodies
CDY1 containing a chromodomain and a histone acetyltransferase catalytic domain. Chromodomain proteins are components of heterochromatin-like complexes and can act as gene repressors. Histone hyperacetylation is thought to facilitate the transition in which protamines replace histones as the major DNA-packaging protein.This gene encodes a protein containing a chromodomain and a histone acetyltransferase catalytic domain. Chromodomain proteins are components of heterochromatin-like complexes and can act as gene repressors. This protein is localized to the nucleus of late spermatids whereMore Product Information