West Nile virus (WNV) matrix protein peptide (15 aa near C-terminus)
Supplier: ProSci Inc.
3431P
PRSI3431PEA 122 EUR
PRSI3431P
West Nile virus (WNV) matrix protein peptide (15 aa near C-terminus)
Proteins and Peptides
West Nile virus peptide is used for blocking the activity of West Nile Virus Matrix Protein antibody.