Anti-CERS5 Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
4697
PRSI4697EA 435 EUR
PRSI4697
Anti-CERS5 Rabbit Polyclonal Antibody
Antibodies
LASS5 Antibody: The LASS (longevity assurance homolog) family members represent a subgroup of the homeobox gene family and are highly conserved from yeasts to mammals. Six members of this family of proteins have been characterized (LASS1-6) and all are involved in ceramide synthesis during cell growth regulation and cancer differentiation. LASS5, also called Trh4, is a 392 amino acid endoplasmic reticulum, multi-pass membrane protein. Functioning as a dihydro-ceramide synthase, LASS5 is involved in the production of sphingolipids containing mainly one fatty acid donor (N-linked palmitoyl-More Product Information
Order Now

SPECIFICATIONS

Antigen symbol CERS5
Antigen name Ceramide Synthase 5
Antigen synonyms Ceramide synthase 5|CerS5|LASS5|LAG1 longevity assurance homolog 5|Trh4
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Immunogen LASS5 antibody was raised against a 18 amino acid synthetic peptide near the amino terminus of the human LASS5.
Purification Affinity chromatography purified via peptide column
Storage buffer PBS containing 0.02% sodium azide
Tested applications E, WB, IHC, IF

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR