Anti-GAPDH Chicken Polyclonal Antibody
Supplier: ProSci Inc.
5003
PRSI5003EA 435 EUR
PRSI5003
Anti-GAPDH Chicken Polyclonal Antibody
Antibodies
GAPDH Antibody: Glyceraldehyde-3-phosphate dehydrogenase (GAPDH) catalyzes the reversible oxidative phosphorylation of glyceraldehyde-3-phosphate in the presence of inorganic phosphate and nicotinamide adenine dinucleotide (NAD), an important energy-yielding step in carbohydrate metabolism. Recent evidence suggests that it also is involved in a number of cellular processes such as membrane fusion, phosphotransferase activity, DNA replication and repair, and nuclear RNA export. GAPDH has also been implicated in playing a role in different pathologies such as cancer progression, apoptosis, andMore Product Information