Anti-GABRG2 Rabbit Polyclonal Antibody
Supplier: ProSci Inc.
55-075
PRSI55-075EA 620 EUR
PRSI55-075
Anti-GABRG2 Rabbit Polyclonal Antibody
Antibodies
This gene encodes a gamma-aminobutyric acid (GABA) receptor. GABA is the major inhibitory neurotransmitter in the mammlian brain, where it acts at GABA-A receptors, which are ligand-gated chloride channels. GABA-A receptors are pentameric, consisting of proteins from several subunit classes: alpha, beta, gamma, delta and rho. Mutations in this gene have been associated with epilepsy and febrile seizures. Multiple transcript variants encoding different isoforms have been identified for this gene.
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.