Human recombinant protocadherin 1 (from cells)

Supplier: ProSci Inc.

91-658
PRSI91-658EA 852 EUR
PRSI91-658
Human recombinant protocadherin 1 (from cells)
Proteins and Peptides

Protocadherin-1, also known as Cadherin-like protein 1, Protocadherin-42 and PCDH1, belongs to the protocadherin subfamily within the cadherin superfamily. PCDH1 contains seven cadherin-like domains, a transmembrane region and a C-terminal cytoplasmic region. PCDH1 can be detected as early as embryonic day 9.5. In early embryogenesis, expression is especially prominent in blood vessels. The tight spatial and temporal regulation of Pcdh1 expression suggests that this protocadherin plays multiple roles not only during development but also in mature tissues and organs. In addition, protocadherin-1 is involved in cell-cell interaction processes and in neural cell adhesion.


  • This recombinant protein can be used for biological assays. For research use only.


Fusion-Tag: C-6 His tag

Order Now

SPECIFICATIONS

Protein/peptide name Protocadherin 1
Environmentally Preferable
Protein synonyms Protocadherin-42|PC42|PCDH1|Protocadherin-1|Cadherin-like Protein 1
Protein/peptide type Recombinant
Species Human
Source Cells
Sequence Thr58-Asn852
Purity > 95% as determined by reducing SDS-PAGE
Formulation Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
Molecular weight 87.3 kD
Storage conditions Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
Tag sequence C-6 His tag
Tested applications Bioassay

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR