Safety helmets, Endurance, PS53
Supplier: Portwest
PS53RBR PS53BKR PS53YER PS53WHR PS53ORR
PRTWPS53RBREA 33 EUR
PRTWPS53RBR PRTWPS53BKR PRTWPS53YER PRTWPS53WHR PRTWPS53ORR
Safety helmets, Endurance, PS53
Head Protection Safety Helmets
Designed to be specially used for working at heights. Light, comfortable and compact with a short peak. ABS shell, textile comfort harness six points, wheel ratchet adjustment, size 52 to 63 cm.
  • Light and robust
  • Specially designed for working at height
  • Electrical insulation up to 1000Vac or 1500Vdc (EN 50365)
  • Six points textile harness
  • Wheel ratchet size adjustment for easy fitting
  • Hi-Visibility colour for visibility in daylight
Comfortable and lightweight, the PPE range will ensure a pleasant wear, even for long periods of use.
Certifications: More Product Information