Long Peak Bump Cap
Supplier: Portwest
PW79ORR
PRTWPW79ORREA 21.1 EUR
PRTWPW79ORR
Long Peak Bump Cap
Head Protection Bump Caps
Modern style bump cap with long peak (70 mm) for ideal protection from glare and provides maximum protection against shock and light.
  • Durable, high-density EVA protects and comforts the wearer
  • Inner ABS shell and soft pad for shock absorption
  • Hook and loop strap for safe and secure fit
  • Padded headband for comfort
  • Retail bag aids presentation for retail sales
Mesh panel on the back, which together with the ventilation holes on the internal shell, provide cooling comfort. Hook and loop adjustable size.