Dissolution baskets, QLA
Supplier: QUALITY LAB ACCESSORIES
BSK010-01 BSK010-HR BSK020-01 BSK020-DK BSK020-HR BSK040-01 BSK040-DK BSKPTD-40 BSKPTH-40 BSKPTV-40 BSKSUP-01 BSKSUP-CA BSKSUP-DK BSKSUP-HR BSKSUP-PT BSKSUP-ST
QLAABSK010-01EA 303 EUR
QLAABSK010-01 QLAABSK010-HR QLAABSK020-01 QLAABSK020-DK QLAABSK020-HR QLAABSK040-01 QLAABSK040-DK QLAABSKPTD-40 QLAABSKPTH-40 QLAABSKPTV-40 QLAABSKSUP-01 QLAABSKSUP-CA QLAABSKSUP-DK QLAABSKSUP-HR QLAABSKSUP-PT QLAABSKSUP-ST
Dissolution baskets, QLA
Dissolution Apparatus
These are clip style and O-ring push-on style baskets for most dissolution baths. All baskets are laser marked with a unique serial number (except suppository baskets).
  • 40 mesh baskets are made from sintered mesh
  • Different mesh sizes are available
The sintering process essentially creates a welded joint at each overlapping wire junction. Sintered baskets are stronger and last longer. They retain original dimensions and geometry and resist wobble. Most conventional baskets are not sintered and weaken over time. Critical basket geometry is verified onMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable