Dissolution calibration tools
Supplier: QUALITY LAB ACCESSORIES
CNTGAG-MI DEPGAG-VK DEPSET-MI DPSPBK-VK DPSPPD-VK LEVEL-UN TACHOM-UN VIBMET-UN WOBMET-MI
QLAACNTGAG-MIEA 2320 EUR
QLAACNTGAG-MI QLAADEPGAG-VK QLAADEPSET-MI QLAADPSPBK-VK QLAADPSPPD-VK QLAALEVEL-UN QLAATACHOM-UN QLAAVIBMET-UN QLAAWOBMET-MI
Dissolution calibration tools
Dissolution Apparatus
These dissolution calibration tools are suitable for most dissolution baths.
  • Mechanical calibration parameters can easily be verified by using calibration tools
  • All calibration tools are serialised
Delivery information: Supplied with NIST traceable calibration certificates and user manuals.