Dishes, crystallising, quartz
Supplier: QUARTZTECHNISCHE WERKSTATTEN
1124
QUTW1124EA 31.2 EUR
QUTW1124
Dishes, crystallising, quartz
Trays/Dishes/Pots Crystallising Dishes
Quartz glass, clear.
  • Extremely low expansion
  • Best possible elastricity and thermal shock resistance
  • High transformation and softening point temperature
  • Low heat conductivity
  • Low electrical conductivity
For a wide range of applications.
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable