Ruthenium(III) chloride oxide, Technical Grade, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Warning

Synonyms: Ruthenium oxychloride, Tetraamminechlorohydroxoruthenium(III) chloride, Ruthenium Red

M-Clarity™ quality level = MQ200

R2751-1G R2751-5G
R2751-1GEA 184 EUR
R2751-1G R2751-5G
Ruthenium(III) chloride oxide, Technical Grade, Sigma-Aldrich®
Ruthenium(III) chloride oxide

Inhibits mitochondrial Ca2+ uniporter; capsaicin antagonist; blocks VR1 vanilloid receptor-coupled ion channels.

Formula: H₄₂Cl₆N₁₄O₂Ru₃
Storage Temperature: Room Temperature
MDL Number: MFCD00011479
CAS Number: 11103-72-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13b337e2c786c7441055418a5a7359c04a063c29&vid=fd77f7c7ac3fbcff563ebcdca3afdbdd02c44a60&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Form) Powder
Solubility (Colour) 10 mg/ml water
Solubility (Method) 10 mg/ml water
Wavelength (1) (Uv) 533 - 534 nm
Extinction (1%/1cm) (1) (Uv) Jun-03
Solvent (UV) Ammonium Acetate 0.1 M
Assay (Icp) 34 - 39 % RUTHENIUM


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR