RNA isolation, viral RNA kit, E.Z.N.A.®
Supplier: OMEGA BIO-TEK
R6874-02 R6874-01 R6874-00
R6874-02EA 748 EUR
R6874-02 R6874-01 R6874-00
RNA isolation, viral RNA kit, E.Z.N.A.®
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The E.Z.N.A.® Viral RNA Kit is designed for the isolation of viral RNA from cell free fluids such as plasma, serum, urine and cell culture supernatant. The procedure completely removes contaminants and enzyme inhibitors making viral RNA isolation fast, convenient and reliable. The kit is also suitable for the isolation of total RNA from cultured cells, tissues and bacteria. RNA purified using the E.Z.N.A.® Viral RNA method is ready for all downstream applications such as RT-PCR.