Reversed phase HPLC columns, PRP-3
Supplier: HAMILTON BONADUZ
79392 79794 79382 79574 79468 79526 79469 79461 79393 79454 79395 79466 79124 79123 79147 79186 79295 79920
R79392EA 1090 EUR
R79392 R79794 R79382 R79574 R79468 R79526 R79469 R79461 R79393 R79454 R79395 R79466 554-1224 554-1223 554-2530 554-2532 HAMI79295 HAMI79920
Reversed phase HPLC columns, PRP-3
Chromatography Columns
PRP-3 polymeric reversed phase columns are ideal for gradient separations of peptides and proteins. The columns are packed with a poly(styrene-divinyl)benzene support.
  • Better sample recovery than silica based columns
  • Excellent durability
  • Last longer than silica based columns
  • Crosslinked to prevent shrinkage or swelling
Since the support is entirely polymeric and there are no C18 groups to hydrolyse, column lifetime is longer. Regardless of whether you use ionpair reagents or high percentage aqueous buffers (80 to 100%) for your separation, PRP-3More Product Information