Cryoboxes
Supplier: RATIOLAB
5220014 5220101 5220010 5220011 5220012 5220013 5220020 5220021 5220022 5220024 5220103 5220100 5220104 5220102
RATI5220014EA 33 EUR
RATI5220014 RATI5220101 RATI5220010 RATI5220011 RATI5220012 RATI5220013 RATI5220020 RATI5220021 RATI5220022 RATI5220024 RATI5220103 RATI5220100 RATI5220104 RATI5220102
Cryoboxes
Boxes Cryoboxes
Cryoboxes are made of polypropylene in natural or other colors. They can be used with sample vessels of differing diameters by removing the grid inserts.
  • Robust hinge with safe snap-on lid
  • Stackable for safe transport
  • Drain bores at the bottom for dew liquids
These cryoboxes are particularly practical and flexible due to their removable inserts. Cleaning of the boxes and the inserts is much simpler than cryoboxes with permanently fitted inserts.
Order Now
SPECIFICATIONS
Environmentally Preferable