Luminescence microplates, 96-well
Supplier: RATIOLAB
6018710
RATI6018710EA 308 EUR
RATI6018710
Luminescence microplates, 96-well
Microplates Immunology Microplates and Strips
These wells are with raised rims for reduced cross-contamination and effective sealing with cover films. Precise design, perfectly shaped for use with automatic systems.
  • SBS standard micro test plate format
  • 8×12 wells alphanumerically coded
  • Optimum space for precise positioning
  • Space-saving and stackable
  • Surface untreated