Diethyl pyrocarbonate

Supplier: G Biosciences
Warning

Synonyms: DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC

RC-042 RC-041
RC-042EA 339 EUR
RC-042 RC-041
Diethyl pyrocarbonate
Diethyl pyrocarbonate
Formula: O(COOC₂H₅)₂
MW: 162.14 g/mol
Boiling Pt: 93
Density: 1.123 g/cm³ (20 °C)
Flash Pt: 69
Storage Temperature: Refrigerator
MDL Number: MFCD00009106
CAS Number: 1609-47-8
EINECS: 216-542-8
Merck Index: 13,08088

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR