X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)

Supplier: G Biosciences

Synonyms: BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

RC-113
RC-113EA 107 EUR
RC-113
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
X-Gal (5-Bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Formula: C₁₄H₁₅BrClNO₆
MW: 408.63 g/mol
Melting Pt: 237…239 °C
Storage Temperature: Freezer
MDL Number: MFCD00005666
CAS Number: 7240-90-6
EINECS: 230-640-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR