Bottles, Narrow Neck, Round, HDPE, UN Approved
Supplier: RIXIUS
3-0600-1000-45
RIXI3-0600-1000-45EA 427 EUR
RIXI3-0600-1000-45
Bottles, Narrow Neck, Round, HDPE, UN Approved
Bottles Plastic Bottles
Narrow-neck bottle made of HDPE, natural.
  • Good chemical resistance
  • Good temperature resistance
  • Robust
Certifications: UN-X approved.