Anti-CECB1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-242
ROCK100-401-242EA 821 EUR
ROCK100-401-242
Anti-CECB1 Rabbit Polyclonal Antibody
Antibodies
Anti-cecropin B is useful for the detection of cecropin B in various immunological assays.  This reagent has specific value for the study of cecropins in transgenic plants. The antibody may also recognize naturally occurring cecropin A.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CECB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Silkworm
Order Now

SPECIFICATIONS

Antigen symbol Cecropin B
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Silkworm
Host Rabbit
Gene ID 693028
Isotype
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Immunogen This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic full-length cecropin B corresponding to a modified sequence from that which is found in silkworm:  Lys-Trp-Lys-Val-Phe-Lys-Lys-Ile-Glu
Format Antiserum
Form Liquid (sterile filtered)
Purification purified by immunoaffinity chromatography
Cross adsorption none
Storage buffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,05% (w/v) Sodium Azide, 50% (v/v) Glycerol
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR