Anti-EIF2AK3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-962
ROCK100-401-962EA 821 EUR
ROCK100-401-962
Anti-EIF2AK3 Rabbit Polyclonal Antibody
Antibodies
This antiserum has been tested for use in western blotting, immunoprecipitation.

Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:500 – 1:3000; Immunoprecipitation: 10-30µl;

Type: Primary
Antigen: EIF2AK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR