Anti-CUL4A Rabbit Monoclonal Antibody (Unconjugated) [clone: 8A3]

Supplier: Rockland Immunochemicals
100-401-A04
ROCK100-401-A04EA 743 EUR
ROCK100-401-A04
Anti-CUL4A Rabbit Monoclonal Antibody (Unconjugated) [clone: 8A3]
Antibodies
This antibody reacts with human Cul4A by western blot and immunoprecipitation.  The antibody immunoprecipitates Protein from cell lysates (using HeLa, U2OS and others).  This antibody also co-immunoprecipitates at least 8 subunits of COP9, DDB1 and CAND1.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;

Type: Primary
Antigen: CUL4A
Clonality: Monoclonal
Clone: 8A3
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol CUL4A
Environmentally Preferable
Antigen name cullin 4A
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID 8451
Isotype
Western blot Yes
ELISA Yes
ImmunoPrecipitation Yes
Epitope N-terminal
Immunogen This antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 1-20 of Human Cul4A (N-terminus) coupled to KLH.
Amino acid number 1 to 20
Format Antiserum
Form Liquid (sterile filtered)
Purification delipidation and defibrination
Concentration 85 mg/ml by Refractometry
Cross adsorption none
Storage buffer 0,01% (w/v) Sodium Azide
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR