Anti-RBX1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-A13
ROCK100-401-A13EA 821 EUR
ROCK100-401-A13
Anti-RBX1 Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with human ROC1 by western blot and immunoprecipitation.  The antibody immunoprecipitates in vitro translated Protein and Protein from overexpressing cell lysates (using HeLa and NIH-3T3, and others).

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;

Type: Primary
Antigen: RBX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol RBX1
Antigen name Ring-box 1, E3 ubiquitin protein ligase
Environmentally Preferable
Antigen synonyms RNF75|BA554C12.1|ROC1
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID 9978
Isotype
Western blot Yes
ELISA Yes
ImmunoPrecipitation Yes
Epitope C-terminal
Immunogen This antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 94-108 of Human ROC1 (C-terminal) coupled to KLH.
Amino acid number 94 to 108
Format Antiserum
Form Liquid (sterile filtered)
Purification delipidation and defibrination
Concentration 85 mg/ml by Refractometry
Cross adsorption none
Storage buffer 0,01% (w/v) Sodium Azide
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR