Anti-Serum Rabbit Monoclonal Antibody (Unconjugated) [clone: 2G9]

Supplier: Rockland Immunochemicals
109-4101
ROCK109-4101EA 504 EUR
ROCK109-4101
Anti-Serum Rabbit Monoclonal Antibody (Unconjugated) [clone: 2G9]
Antibodies
Anti-human serum antibody is suitable for ELISA and western blotting. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum and human Serum.

Recommended Dilutions:
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: Serum
Clonality: Monoclonal
Clone: 2G9
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR