Anti-lambda light chainlambda light chain Rabbit Polyclonal Antibody (Texas Red®)

Supplier: Rockland Immunochemicals
610-4911
ROCK610-4911EA 436 EUR
ROCK610-4911
Anti-lambda light chainlambda light chain Rabbit Polyclonal Antibody (Texas Red®)
Antibodies
Secondary Rabbit Anti-λ (lambda chain) Reacts with Mouse

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgL
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: Light Chain
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR