Anti-IgG (H+L) Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
712-1633
ROCK712-1633EA 457 EUR
ROCK712-1633
Anti-IgG (H+L) Goat Polyclonal Antibody (Biotin)
Antibodies
This product has been assayed against 1.0 µg of Rat IgG in a standard capture ELISA using Peroxidase Conjugated Streptavidin and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 500 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

SPECIFICATIONS

Antigen symbol IgG (H+L)
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Conjugation Biotin
Reactivity Rat
Host Goat
Isotype
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Gel shift Yes
Flow cytometry Yes
ChIP Yes
Neutralisation Yes
Antibody fragment F(ab')2
Immunogen Rat IgG whole molecule
Format Antibody fraction
Form Lyophilized
Purification purified by immunoaffinity chromatography
Concentration 1,0 mg/ml by UV absorbance at 280 nm
Storage buffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR