Polyethylene glycol 6,000

Supplier: Roth Carl

0158.1 0158.2 0158.3 0158.4
ROTH0158.1EA 0 EUR
ROTH0158.1 ROTH0158.2 ROTH0158.3 ROTH0158.4
Polyethylene glycol 6,000
Polyethylene glycol 6,000
Formula: H(OCH₂CH₂)nOH
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 55…62 °C
Density: 1,13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD01779614
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR