Manganese(II) acetate tetrahydrate

Supplier: Roth Carl

Synonyms: Manganese acetate tetrahydrate, Manganese diacetate tetrahydrate, Acetic acid manganese (II) salt tetrahydrate

0277.1 0277.2 0277.3
ROTH0277.1EA 0 EUR
ROTH0277.1 ROTH0277.2 ROTH0277.3
Manganese(II) acetate tetrahydrate
Manganese (II) acetate tetrahydrate
Formula: (H₃CCOO)₂Mn·4H₂O
Melting Pt: 300 °C
Density: 1,58 g/cm³ (20 °C)
MDL Number: MFCD00062552
CAS Number: 6156-78-1
EINECS: 211-334-3
Merck Index: 13,05746

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR