X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)

Supplier: Roth Carl

2315.2 2315.1 2315.3 2315.4
ROTH2315.2EA 0 EUR
ROTH2315.2 ROTH2315.1 ROTH2315.3 ROTH2315.4
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
X-Gal (5-Bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Formula: C₁₄H₁₅BrClNO₆
Melting Pt: 237…239 °C
Storage Temperature: Freezer
MDL Number: MFCD00005666
CAS Number: 7240-90-6
EINECS: 230-640-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR