Isovaleraldehyde

Supplier: Roth Carl

Synonyms: 3-Methylbutyraldehyde

3035.1
ROTH3035.1EA 0 EUR
ROTH3035.1
Isovaleraldehyde
Isovaleraldehyde
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 90
Melting Pt: –60 °C
Density: 0.8 g/cm³ (20 °C)
Flash Pt: 0
Storage Temperature: Refrigerator
MDL Number: MFCD00007014
CAS Number: 590-86-3
EINECS: 209-691-5
UN: 2058
ADR: 3 II
Merck Index: 13,05250

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR