1-Octanol

Supplier: Roth Carl
Warning

Synonyms: n-Octanol

3182.1
ROTH3182.1EA 148 EUR
ROTH3182.1
1-Octanol
1-Octanol
Formula: H₃C(CH₂)₆CH₂OH
MW: 130,23 g/mol
Boiling Pt: 195 °C (1013 hPa)
Melting Pt: –16 °C
Density: 0,8246 g/cm³ (20 °C)
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00002988
CAS Number: 111-87-5
EINECS: 203-917-6
Merck Index: 13,06784
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7db4a6be3aedee4af43781ec6deb31631bc376c1&vid=80cb13f618a2d3181daf8310324993f1f7c69fb2&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR