Petroleum spirit, 80…110 °C

Supplier: Roth Carl
Danger

3259.1 3259.2
ROTH3259.1EA 0 EUR
ROTH3259.1 ROTH3259.2
Petroleum spirit, 80…110 °C
Petroleum spirit
Boiling Pt: 80…110 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR