Dextrin

Supplier: Roth Carl

Synonyms: Dextrin

3488.1
ROTH3488.1EA 0 EUR
ROTH3488.1
Dextrin
Dextrin
Formula: (C₆H₁₂O₆)n
MW: 180.16 g/mol
Density: 1.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081554
CAS Number: 9004-53-9
EINECS: 232-675-4
Merck Index: 13,02971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR