p-Anisic acid

Supplier: Roth Carl

Synonyms: 4-Methoxybenzoic acid, 4-Anisic acid

3631.1
ROTH3631.1EA 0 EUR
ROTH3631.1
p-Anisic acid
p-Anisic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 275
Melting Pt: 182…185 °C
Density: ∼1.385 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002542
CAS Number: 100-09-4
EINECS: 202-818-5
Merck Index: 13,00669

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR