tetra-Chloroauric(III) acid trihydrate

Supplier: Roth Carl
Danger

Synonyms: Gold(III) chloride trihydrate, Gold Chloride hydrate, Hydrogen tetrachloroaurate(III) trihydrate, Hydrogen tetrachloroaurate (III) trihydrate, tetra-Chloroauric acid trihydrate

3867.1 3867.2
ROTH3867.1EA 0 EUR
ROTH3867.1 ROTH3867.2
tetra-Chloroauric(III) acid trihydrate
tetra-Chloroauric (III) acid trihydrate
Formula: HAuCl₄·3H₂O
MW: 393.83 g/mol
Melting Pt: 30
Density: 3.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149904
CAS Number: 16961-25-4
EINECS: 240-948-4
UN: 3260
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR