L(+)-Isoleucine

Supplier: Roth Carl

3922.1 3922.2 3922.3
ROTH3922.1EA 0 EUR
ROTH3922.1 ROTH3922.2 ROTH3922.3
L(+)-Isoleucine
L(+)-Isoleucine
Formula: H₃CCH₂CH(CH₃)CH(NH₂)COOH
Melting Pt: 279…280 °C
Storage Temperature: Ambient
MDL Number: MFCD00064222
CAS Number: 73-32-5
EINECS: 200-798-2
Merck Index: 14,05179

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR