β-Ionone

Supplier: Roth Carl

3951.1
ROTH3951.1EA 0 EUR
ROTH3951.1
β-Ionone
β-Ionone
Formula: C₁₃H₂₀O
MDL Number: MFCD00001549
CAS Number: 79-77-6
EINECS: 201-224-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR