Pyrocatechol

Supplier: Roth Carl
Warning

Synonyms: o-Dihydroxybenzene, Catechol, 1,2-Dihydroxybenzene

4249.1
ROTH4249.1EA 0 EUR
ROTH4249.1
Pyrocatechol
Pyrocatechol
Formula: C₆H₆O₂
MW: 110.11 g/mol
Boiling Pt: 245
Melting Pt: 103…105 °C
Density: 1.0548 g/cm³ (20 °C)
Flash Pt: 127
Storage Temperature: Ambient
MDL Number: MFCD00002188
CAS Number: 120-80-9
EINECS: 204-427-5
UN: 2811
ADR: 6.1 III
Merck Index: 13,08089

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR