Lead(II) oxide

Supplier: Roth Carl
Danger

Synonyms: Lead monoxide

4456.1
ROTH4456.1EA 0 EUR
ROTH4456.1
Lead(II) oxide
Lead (II) oxide
Formula: PbO
MW: 223.2 g/mol
Boiling Pt: 1470
Melting Pt: 888
Density: 9.5 g/cm³ (20 °C)
MDL Number: MFCD00011164
CAS Number: 1317-36-8
EINECS: 215-267-0
UN: 2291
ADR: 6.1 III
Merck Index: 13,05431

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR