Iron(III) oxide

Supplier: Roth Carl
Warning

Synonyms: Ferric oxide, Ferric Oxide, (iron(III) oxide), Diiron trioxide

4463.1
ROTH4463.1EA 0 EUR
ROTH4463.1
Iron(III) oxide
Iron (III) oxide
Formula: Fe₂O₃
Melting Pt: 1538 °C
Density: 5,24 g/cm³ (20 °C)
MDL Number: MFCD00011008
CAS Number: 1309-37-1
EINECS: 215-168-2
Merck Index: 13,04058

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR