Acetic anhydride

Supplier: Roth Carl
Danger

4483.1 4483.2
ROTH4483.1EA 0 EUR
ROTH4483.1 ROTH4483.2
Acetic anhydride
Acetic anhydride
Formula: (CH₃CO)₂O
Boiling Pt: 139,9 °C (1013 hPa)
Melting Pt: –73 °C
Density: 1,08 g/cm³ (20 °C)
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00008705
CAS Number: 108-24-7
EINECS: 203-564-8
UN: 1715
ADR: 8,II
REACH: 01-2119486470-36
Merck Index: 13,00057
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f107a66d926eb36bee4d132880765bc7fc2db7c8&vid=1c6b77698bd69c2f562bcda9bf02b8dc694a0d16&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR