p-Toluenesulphonic acid monohydrate

Supplier: Roth Carl
Warning

Synonyms: 4-Methylbenzenesulphonic acid hydrate, Toluene-4-sulphonic acid monohydrate

4796.1 4796.2
ROTH4796.1EA 40.2 EUR
ROTH4796.1 ROTH4796.2
p-Toluenesulphonic acid monohydrate
p-Toluenesulphonic acid monohydrate
Formula: C₇H₈O₃S·1H₂O
MW: 190.22 g/mol
Melting Pt: 102…104 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 180
Storage Temperature: Ambient
MDL Number: MFCD00142137
CAS Number: 6192-52-5
EINECS: 203-180-0
UN: 2585
ADR: 8 III
Merck Index: 12,09671

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR