Cinnamyl alcohol

Supplier: Roth Carl

4905.1
ROTH4905.1EA 23.5 EUR
ROTH4905.1
Cinnamyl alcohol
Cinnamyl alcohol
Formula: C₉H₁₀O
Boiling Pt: 256 °C (1013 hPa)
Melting Pt: 31…34 °C
Density: 1,04 g/cm³ (25 °C)
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00002921
CAS Number: 104-54-1
EINECS: 203-212-3
Merck Index: 13,02325

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR