Digitonin

Supplier: Roth Carl

4946.1 4946.2
ROTH4946.1EA 0 EUR
ROTH4946.1 ROTH4946.2
Digitonin
Digitonin
Formula: C₅₆H₉₂O₂₉
MDL Number: MFCD00077729
CAS Number: 11024-24-1
EINECS: 234-255-6
UN: 3462
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR