Acetone ≥99.5% (by GC) for synthesis

Supplier: Roth Carl
Danger

5025.3 5025.1 5025.2 5025.4 5025.5 5025.6
ROTH5025.3EA 0 EUR
ROTH5025.3 ROTH5025.1 ROTH5025.2 ROTH5025.4 ROTH5025.5 ROTH5025.6
Acetone ≥99.5% (by GC) for synthesis
Acetone
Formula: CH₃COCH₃
Boiling Pt: 56,2 °C (1013 hPa)
Melting Pt: –95,4 °C
Density: 0,792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance clear, colourless liquid
Assay (GC) ≥99,5 %
Density (20 °C) 0,788-0,793 g/cm³
Water (KF) ≤0,3 %
Refractive index (20 °C) 1,358-1,360


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR